Benzo[b]furan-3-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Benzofuran-3-ylboronic acid

FA-2484-1G FA-2484-250M
COBBFA-2484-1GEA 153 EUR
COBBFA-2484-1G COBBFA-2484-250M
Benzo[b]furan-3-boronsäure
Benzo[b]furan-3-boronsäure
Formel: C₈H₇BO₃
Molecular Weight: 161,95 g/mol
MDL: MFCD06801687
CAS-Nummer: 317830-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR