4-Dibenzofuranboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid

BB-8305-5G BB-8305-1G
COBBBB-8305-5GEA 50.9 EUR
COBBBB-8305-5G COBBBB-8305-1G
4-Dibenzofuranboronsäure
4-Dibenzofuranboronic acid
Formel: C₁₂H₉BO₃
Molecular Weight: 212,01 g/mol
MDL: MFCD00092336
CAS-Nummer: 100124-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR