2-Brom-5-formylthiazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromothiazole-5-carboxaldehyde

HI-1125-5G HI-1125-25G HI-1125-1G
COBBHI-1125-5GEA 223 EUR
COBBHI-1125-5G COBBHI-1125-25G COBBHI-1125-1G
2-Brom-5-formylthiazol
2-Brom-5-formylthiazol
Formel: C₄H₂BrNOS
Molecular Weight: 192,03 g/mol
MDL: MFCD03788567
CAS-Nummer: 464192-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR