2-Brom-4-methylthiazol-5-carbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-Brom-4-methyl-1,3-thiazol-5-carbonsäure

MO00751DA MO00751EA
MAYBMO00751DAEA 0 EUR
MAYBMO00751DA MAYBMO00751EA
2-Brom-4-methylthiazol-5-carbonsäure
2-Brom-4-methylthiazol-5-carbonsäure
Formel: C₅H₄BrNO₂S
Molecular Weight: 222.06 g/mol
MDL: MFCD03822128
CAS-Nummer: 40003-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR