2-Brom-4-methylthiazol-5-carbonsäure ≥96%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Brom-4-methyl-1,3-thiazol-5-carbonsäure

M-Clarity™ Qualitätsstufe = MQ100

646008-5G 646008-1G
SIAL646008-5GEA 266 EUR
SIAL646008-5G SIAL646008-1G
2-Brom-4-methylthiazol-5-carbonsäure ≥96%, Sigma-Aldrich®
2-Brom-4-methylthiazol-5-carbonsäure
Formel: C₅H₄BrNO₂S
Molecular Weight: 222.06 g/mol
MDL: MFCD09026784
CAS-Nummer: 40003-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR