2-Amino-4,5-dimethylthiazol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,5-Dimethylthiazol-2-amine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

174408-50G 174408-10G
SIAL174408-50GEA 198 EUR
SIAL174408-50G SIAL174408-10G
2-Amino-4,5-dimethylthiazol Hydrochlorid, Sigma-Aldrich®
2-Amino-4,5-dimethylthiazol Hydrochlorid
Formel: C₅H₉ClN₂S
Molecular Weight: 164,66 g/mol
MDL: MFCD00012711
CAS-Nummer: 71574-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR