trans-4-Isopropyl-β-nitrostyrol
Lieferant: Thermo Scientific
Synonyme:
4-Isopropyl-β-nitrostyrene
S14725.1
S14725.10
S14725.50
MAYBS14725.1EA
0
EUR
MAYBS14725.1
MAYBS14725.10
MAYBS14725.50
trans-4-Isopropyl-β-nitrostyrol
trans-4-Isopropyl-β-nitrostyrol
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191.23 g/mol |
MDL:
MFCD00085100 CAS-Nummer: 42139-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...