trans-4-Methyl-β-nitrostyrol

Lieferant: Thermo Scientific

S01418.10 S01418.50
MAYBS01418.10EA 0 EUR
MAYBS01418.10 MAYBS01418.50
trans-4-Methyl-β-nitrostyrol
trans-4-Methyl-β-nitrostyrol
Formel: C₉H₉NO₂
MDL: MFCD00024828
CAS-Nummer: 5153-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR