4-Fluorbenzylidenaceton

Lieferant: Thermo Scientific

Synonyme: 1-(4-Fluorophenyl)but-1-en-3-one

CD11350.250 CD11350.1
MAYBCD11350.250EA 0 EUR
MAYBCD11350.250 MAYBCD11350.1
4-Fluorbenzylidenaceton
4-Fluorbenzylidenaceton
Formel: C₁₀H₉FO
MDL: MFCD00205144
CAS-Nummer: 1611-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR