1,2-Dibrompentafluorpropyl-2,2,2,3,3-pentafluorpropylether

Lieferant: FLUOROCHEM

010286-1G 010286-5G
FLUO010286-1GEA 60.9 EUR
FLUO010286-1G FLUO010286-5G
1,2-Dibrompentafluorpropyl-2,2,2,3,3-pentafluorpropylether
1,2-Dibrompentafluorpropyl-2,2,2,3,3-pentafluorpropylether
Formel: C₆H₂Br₂F₁₀O
MDL: MFCD03094147
CAS-Nummer: 396716-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR