1,2-Dibrompentafluorpropyl-2,2,3,3-tetrafluorpropylether
Lieferant: FLUOROCHEM
010287-1G
010287-5G
FLUO010287-1GEA
57.9
EUR
FLUO010287-1G
FLUO010287-5G
1,2-Dibrompentafluorpropyl-2,2,3,3-tetrafluorpropylether
1,2-Dibrompentafluorpropyl-2,2,3,3-tetrafluorpropylether
Formel:
C₆H₃Br₂F₉O |
MDL:
MFCD03094408 CAS-Nummer: 396716-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...