2-(4-Formylphenoxy)-2-phenylessigsäure
Lieferant: BIONET RESEARCH
5Y-0807.5
5Y-0807.500
5Y-0807.10
5Y-0807.1
BIOT5Y-0807.5EA
0
EUR
BIOT5Y-0807.5
BIOT5Y-0807.500
BIOT5Y-0807.10
BIOT5Y-0807.1
2-(4-Formylphenoxy)-2-phenylessigsäure
2-(4-Formylphenoxy)-2-phenylessigsäure
Formel:
C₁₅H₁₂O₄ |
CAS-Nummer:
480994-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...