2-(4-Acetylphenoxy)-2-phenylessigsäure

Lieferant: BIONET RESEARCH

6Y-0808.500 6Y-0808.5 6Y-0808.10 6Y-0808.1
BIOT6Y-0808.500EA 0 EUR
BIOT6Y-0808.500 BIOT6Y-0808.5 BIOT6Y-0808.10 BIOT6Y-0808.1
2-(4-Acetylphenoxy)-2-phenylessigsäure
2-(4-Acetylphenoxy)-2-phenylessigsäure
Formel: C₁₆H₁₄O₄
CAS-Nummer: 885949-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR