Ethyl-3-aminocrotonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 3-aminobut-2-enoate

M-Clarity™ Qualitätsstufe = MQ200

E10807-100G E10807-500G
SIALE10807-100GEA 49.8 EUR
SIALE10807-100G SIALE10807-500G
Ethyl-3-aminocrotonat, Sigma-Aldrich®
Ethyl-3-aminocrotonat
Formel: C₆H₁₁NO₂
Molecular Weight: 129,16 g/mol
MDL: MFCD00008073
CAS-Nummer: 7318-00-5
EINECS: 230-782-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR