Methyl-3-aminocrotonat 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate
156790050
156791000
156795000
ACRO156790050EA
23.4
EUR
ACRO156790050
ACRO156791000
ACRO156795000
Methyl-3-aminocrotonat 97%
Methyl-3-aminocrotonat
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Schmelzpunkt: 81…83 °C Flammpunkt: 91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008072 CAS-Nummer: 14205-39-1 EINECS: 238-056-5 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 81 to 85 °C |
Titration with HClO4 | ≥96.0 % |
Sulfate (SO4) | ≤0.005 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Ammonia (NH3) | ≤0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...