Methyl-3-aminocrotonat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate

B24953.22 B24953.36
B24953.22EA 29.9 EUR
B24953.22 B24953.36
Methyl-3-aminocrotonat 97%
Methyl-3-aminocrotonat

Purity: 97% CAS No.: 14205-39-1 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Flash Point: 91°(195°F) Melting Point: 81-85° Boiling Point: 122°/42mm Beilstein Reference: 956592 MDL No.: MFCD00008072

Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Siedepunkt: 122 °C (42 mmHg)
Schmelzpunkt: 81…85 °C
Flammpunkt: 91 °C (195 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008072
CAS-Nummer: 14205-39-1
EINECS: 238-056-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR