Methyl-3-aminocrotonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate

M-Clarity™ Qualitätsstufe = MQ200

129712-100G 129712-500G
SIAL129712-100GEA 45.4 EUR
SIAL129712-100G SIAL129712-500G
Methyl-3-aminocrotonat, Sigma-Aldrich®
Methyl-3-aminocrotonat
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
MDL: MFCD00008072
CAS-Nummer: 14205-39-1
EINECS: 238-056-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR