Methyl-3-aminocrotonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 3-Amino-2-butenoate, 3-Aminocrotonic acid Methyl Ester, Methyl 3-aminobut-2-enoate, 3-Amino-2-butenoic acid Methyl Ester, Methyl 3-Aminocrotonate
M-Clarity™ Qualitätsstufe = MQ200
688894-10KG
688894-1KG
688894-50KG
SIAL688894-10KGEA
1230
EUR
SIAL688894-10KG
SIAL688894-1KG
SIAL688894-50KG
Methyl-3-aminocrotonat, Sigma-Aldrich®
Methyl-3-aminocrotonat
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol |
MDL:
MFCD00008072 CAS-Nummer: 14205-39-1 EINECS: 238-056-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...