Ethyl-3-amino-4,4,4-trifluorcrotonat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 3-amino-4,4,4-trifluorobut-2-enoate
001495-10G
001495-50G
001495-250G
FLUO001495-10GEA
36.9
EUR
FLUO001495-10G
FLUO001495-50G
FLUO001495-250G
Ethyl-3-amino-4,4,4-trifluorcrotonat
Ethyl-3-amino-4,4,4-trifluorcrotonat
Formel:
C₆H₈F₃NO₂ Molecular Weight: 183,13 g/mol |
MDL:
MFCD00068195 CAS-Nummer: 372-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...