(4-Acetamidophenyl)boronsäure

Lieferant: FLUOROCHEM

010749.2G 010749-5G 010749-1G
FLUO010749.2GEA 170 EUR
FLUO010749.2G FLUO010749-5G FLUO010749-1G
(4-Acetamidophenyl)boronsäure
(4-Acetamidophenyl)boronsäure
Formel: C₈H₁₀BNO₃
MDL: MFCD02179451
CAS-Nummer: 101251-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR