Methyl-2-amino-5-brombenzoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 2-amino-5-bromobenzoate

A16666.22 A16666.06 A16666.14
A16666.22EA 276 EUR
A16666.22 A16666.06 A16666.14
Methyl-2-amino-5-brombenzoat 98%
Methyl-2-amino-5-brombenzoat

Purity: 98% CAS No.: 52727-57-8 Molecular Formula: C8H8BrNO2 Formula Weight: 230.06 Melting Point: 71-73° Beilstein Reference: 972720

Formel: C₈H₈BrNO₂
Molecular Weight: 230.06 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00144761
CAS-Nummer: 52727-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR