4-(4-Bromphenoxy)anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

695394-25G 695394-5G
SIAL695394-25GEA 468 EUR
SIAL695394-25G SIAL695394-5G
4-(4-Bromphenoxy)anilin, Sigma-Aldrich®
4-(4-Bromphenoxy)anilin
Formel: C₁₂H₁₀BrNO
CAS-Nummer: 31465-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR