Methyl-2-amino-4-chlorbenzoat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-amino-4-chlorobenzoate

L11125.09 L11125.18
L11125.09EA 48.9 EUR
L11125.09 L11125.18
Methyl-2-amino-4-chlorbenzoat 98%
Methyl-2-amino-4-chlorbenzoat

Purity: 98% CAS No.: 5900-58-3 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 Melting Point: 66-68° Beilstein Reference: 1072895

Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017568
CAS-Nummer: 5900-58-3
EINECS: 227-593-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR