Methyl-2-amino-5-chlorbenzoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-amino-5-chlorobenzoate

L20187.06 L20187.14
L20187.06EA 76.4 EUR
L20187.06 L20187.14
Methyl-2-amino-5-chlorbenzoat ≥98%
Methyl-2-amino-5-chlorbenzoat

Purity: 98+% CAS No.: 5202-89-1 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 Melting Point: 67-70° Boiling Point: 168-170°/1mm MDL No.: MFCD00007837

Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Siedepunkt: 168…170 °C (1 mmHg)
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007837
CAS-Nummer: 5202-89-1
EINECS: 225-992-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR