4-(4-Chlorphenoxy)anilin

Lieferant: Thermo Scientific

Synonyme: 4-Amino-4'-chlorodiphenyl ether

SPB06381.1 SPB06381.10
MAYBSPB06381.1EA 0 EUR
MAYBSPB06381.1 MAYBSPB06381.10
4-(4-Chlorphenoxy)anilin
4-(4-Chlorphenoxy)anilin
Formel: C₁₂H₁₀ClNO
Molecular Weight: 219,67 g/mol
MDL: MFCD00043925
CAS-Nummer: 101-79-1
EINECS: 202-976-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR