Ethyl-2-amino-5-nitrobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 2-amino-5-nitrobenzoate

DP01680.250 DP01680.1
CHMPDP01680.250EA 0 EUR
CHMPDP01680.250 MAYBDP01680.1
Ethyl-2-amino-5-nitrobenzoat
Ethyl-2-amino-5-nitrobenzoat
Formel: C₈H₈N₂O₄
Molecular Weight: 196.1601 g/mol
MDL: MFCD00278157
CAS-Nummer: 3816-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR