Methyl-2-amino-5-fluorbenzoat 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 2-Amino-5-fluorobenzoate, 5-Fluoroanthranilic acid methyll ester, Methyl 5-Fluoroanthranilate, 2-Amino-5-fluorobenzoic acid methyll ester, Methyl 2-amino-5-fluorobenzoate

L19778.06 L19778.14
L19778.06EA 58 EUR
L19778.06 L19778.14
Methyl-2-amino-5-fluorbenzoat 98%
Methyl-2-amino-5-fluorbenzoat

Purity: 98% CAS No.: 319-24-4 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Flash Point: 132°(269°F) Melting Point: 21-25° Boiling Point: 100-102°/0.5mm Beilstein Reference: 2803210 MDL No.: MFCD03791308

Formel: C₈H₈FNO₂
Molecular Weight: 169.16 g/mol
Siedepunkt: 100…102 °C (0.5 mmHg)
Schmelzpunkt: 21…25 °C
Dichte: 1.260
Flammpunkt: 132 °C (269 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03791308
CAS-Nummer: 319-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR