Methyl-2-amino-5-fluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: Methyl 2-Amino-5-fluorobenzoate, 5-Fluoroanthranilic acid methyll ester, Methyl 5-Fluoroanthranilate, 2-Amino-5-fluorobenzoic acid methyll ester, Methyl 2-amino-5-fluorobenzoate

013960-25G 013960-5G
FLUO013960-25GEA 146 EUR
FLUO013960-25G FLUO013960-5G
Methyl-2-amino-5-fluorbenzoat
Methyl-2-amino-5-fluorbenzoat
Formel: C₈H₈FNO₂
Molecular Weight: 169,16 g/mol
MDL: MFCD03791308
CAS-Nummer: 319-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR