4-(3-(Trifluormethyl)phenoxy)anilin

Lieferant: FLUOROCHEM

011240-1G 011240-5G
FLUO011240-1GEA 0 EUR
FLUO011240-1G FLUO011240-5G
4-(3-(Trifluormethyl)phenoxy)anilin
4-(3-(Trifluormethyl)phenoxy)anilin
Formel: C₁₃H₁₀F₃NO
CAS-Nummer: 41605-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR