4-(3-(Trifluormethyl)phenoxy)anilin

Lieferant: MENAI

NC2216.1 NC2216.2 NC2216.5 NC2216.10
MENANC2216.1EA 0 EUR
MENANC2216.1 MENANC2216.2 MENANC2216.5 MENANC2216.10
4-(3-(Trifluormethyl)phenoxy)anilin
4-(3-(Trifluormethyl)phenoxy)anilin
Formel: C₁₃H₁₀F₃NO
CAS-Nummer: 41605-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR