4-(4-(Trifluormethyl)phenoxy)anilin

Lieferant: FLUOROCHEM

011239-1G 011239-5G
FLUO011239-1GEA 0 EUR
FLUO011239-1G FLUO011239-5G
4-(4-(Trifluormethyl)phenoxy)anilin
4-(4-(Trifluormethyl)phenoxy)anilin
Formel: C₁₃H₁₀F₃NO
Molecular Weight: 253.220 g/mol
MDL: MFCD00170930
CAS-Nummer: 57478-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR