4-(4-(Trifluormethyl)phenoxy)anilin

Lieferant: MENAI

E1750.2 E1750.5 E1750.10 E1750.1
MENAE1750.2EA 0 EUR
MENAE1750.2 MENAE1750.5 MENAE1750.10 MENAE1750.1
4-(4-(Trifluormethyl)phenoxy)anilin
4-(4-(Trifluormethyl)phenoxy)anilin
Formel: C₁₃H₁₀F₃NO
Molecular Weight: 253.220 g/mol
MDL: MFCD00170930
CAS-Nummer: 57478-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR