4-[(Trifluormethyl)thio]anilin

Lieferant: CHEMPUR

CFF1301. FL4015.5 FL4015.10
CHMPCFF1301.EA 0 EUR
CHMPCFF1301. CHMPFL4015.5 CHMPFL4015.10
4-[(Trifluormethyl)thio]anilin
4-[(Trifluormethyl)thio]anilin
Formel: C₇H₆F₃NS
MDL: MFCD00040926
CAS-Nummer: 372-16-7
EINECS: 206-744-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR