4-[(Trifluormethyl)thio]anilin

Lieferant: CHEMPUR

Synonyme: 4-((Trifluoromethyl)thio)aniline

TL00414.250 TL00414.1
CHMPTL00414.250EA 0 EUR
CHMPTL00414.250 MAYBTL00414.1
4-[(Trifluormethyl)thio]anilin
4-[(Trifluormethyl)thio]anilin
Formel: C₇H₆F₃NS
Molecular Weight: 193.19 g/mol
MDL: MFCD00040926
CAS-Nummer: 372-16-7
EINECS: 206-744-4
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR