4-[(Trifluormethyl)thio]anilin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-((Trifluoromethyl)thio)aniline
M-Clarity™ Qualitätsstufe = MQ100
470074-1G
470074-5G
SIAL470074-1GEA
51.3
EUR
SIAL470074-1G
SIAL470074-5G
4-[(Trifluormethyl)thio]anilin, Sigma-Aldrich®
4-[(Trifluormethyl)thio]anilin
Formel:
C₇H₆F₃NS Molecular Weight: 193,19 g/mol |
MDL:
MFCD00040926 CAS-Nummer: 372-16-7 EINECS: 206-744-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...