4-[(Trifluormethyl)sulfonyl]anilin

Lieferant: CHEMPUR

FL7802.1 FL7802.5
CHMPFL7802.1EA 0 EUR
CHMPFL7802.1 CHMPFL7802.5
4-[(Trifluormethyl)sulfonyl]anilin
4-[(Trifluormethyl)sulfonyl]anilin
Formel: C₇H₆F₃NO₂S
CAS-Nummer: 473-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR