4-[(Trifluormethyl)sulfonyl]anilin

Lieferant: CHEMPUR

BTB13853.250 BTB13853.1
CHMPBTB13853.250EA 0 EUR
CHMPBTB13853.250 MAYBBTB13853.1
4-[(Trifluormethyl)sulfonyl]anilin
4-[(Trifluormethyl)sulfonyl]anilin
Formel: C₇H₆F₃NO₂S
CAS-Nummer: 473-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR