4-[(Trifluormethyl)sulfonyl]anilin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
481920-1G
481920-500MG
SIAL481920-1GEA
162
EUR
SIAL481920-1G
SIAL481920-500MG
4-[(Trifluormethyl)sulfonyl]anilin, Sigma-Aldrich®
4-[(Trifluormethyl)sulfonyl]anilin
Formel:
C₇H₆F₃NO₂S |
CAS-Nummer:
473-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...