Ethyl-2-amino-5-iodbenzoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-amino-5-iodobenzoate

B25484.22 B25484.14
B25484.22EA 78 EUR
B25484.22 B25484.14
Ethyl-2-amino-5-iodbenzoat ≥98%
Ethyl-2-amino-5-iodbenzoat

Purity: 98+% CAS No.: 268568-11-2 Molecular Formula: C9H10INO2 Formula Weight: 291.09 Melting Point: 69-71° MDL No.: MFCD04038940

Formel: C₉H₁₀INO₂
Molecular Weight: 291,09 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038940
CAS-Nummer: 268568-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR