trans-4-Brom-β-nitrostyrol

Lieferant: CHEMPUR

Synonyme: (E)-1-Bromo-4-(2-nitrovinyl)benzene

JFD00653.250 JFD00653.1
CHMPJFD00653.250EA 0 EUR
CHMPJFD00653.250 MAYBJFD00653.1
trans-4-Brom-β-nitrostyrol
trans-4-Brom-β-nitrostyrol
Formel: C₈H₆BrNO₂
Molecular Weight: 228.0427 g/mol
MDL: MFCD00191856
CAS-Nummer: 5153-71-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR