trans-3,4-Dichlor-β-nitrostyrol

Lieferant: Thermo Scientific

Synonyme: 3,4-Dichloro-β-nitrostyrene, 1,2-Dichloro-4-[(E)-2-nitroethenyl]benzene

S12121.1 S12121.10 S12121.50
MAYBS12121.1EA 0 EUR
MAYBS12121.1 MAYBS12121.10 MAYBS12121.50
trans-3,4-Dichlor-β-nitrostyrol
trans-3,4-Dichlor-β-nitrostyrol
Formel: C₈H₅Cl₂NO₂
MDL: MFCD00052834
CAS-Nummer: 18984-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR