trans-2,6-Dichlor-β-nitrostyrol
Lieferant: Thermo Scientific
Synonyme:
2,6-Dichloro-β-nitrostyrene
S01398.1
S01398EA
S01398.50
MAYBS01398.1EA
0
EUR
MAYBS01398.1
MAYBS01398EA
MAYBS01398.50
trans-2,6-Dichlor-β-nitrostyrol
trans-2,6-Dichlor-β-nitrostyrol
Formel:
C₈H₅Cl₂NO₂ Molecular Weight: 218,04 g/mol |
MDL:
MFCD00024821 CAS-Nummer: 22482-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...