2,4-Dichloro-1-(2-nitrovinyl)benzene, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-2,4-Dichloro-β-nitrostyrene

M-Clarity™ Qualitätsstufe = MQ100

642169-1G 642169-5G
SIAL642169-1GEA 51.5 EUR
SIAL642169-1G SIAL642169-5G
2,4-Dichloro-1-(2-nitrovinyl)benzene, Sigma-Aldrich®
2,4-Dichloro-1-(2-nitrovinyl)benzene
Formel: C₈H₅Cl₂NO₂
Molecular Weight: 218,037 g/mol
MDL: MFCD00053063
CAS-Nummer: 18984-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR