trans-2-Chlor-β-nitrostyrol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloro-β-nitrostyrene, 1-(2-Chlorophenyl)-2-nitroethylene

A14882.06 A14882.14
A14882.06EA 58.6 EUR
A14882.06 A14882.14
trans-2-Chlor-β-nitrostyrol ≥98%
trans-2-Chlor-β-nitrostyrol

Purity: 98% CAS No.: 3156-34-1 Molecular Formula: C8H6ClNO2 Formula Weight: 183.60 Melting Point: 46-49° Beilstein Reference: 2328095

Formel: C₈H₆ClNO₂
Schmelzpunkt: 46…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024820
CAS-Nummer: 3156-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR