3-Nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Nitro-3-vinylbenzene

M-Clarity™ Qualitätsstufe = MQ100

N26601-10G N26601-2.5G
SIALN26601-10GEA 209 EUR
SIALN26601-10G SIALN26601-2.5G
3-Nitrostyrol, Sigma-Aldrich®
3-Nitrostyrol
Formel: C₈H₇NO₂
MDL: MFCD00007276
CAS-Nummer: 586-39-0
EINECS: 209-575-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR