4-Nitrostyrol

Lieferant: BIONET RESEARCH

Synonyme: 1-Nitro-4-vinylbenzene, 1-Ethenyl-4-nitrobenzene

9T-0616.500 9T-0616.1 9T-0616.5 9T-0616.10
BIOT9T-0616.500EA 0 EUR
BIOT9T-0616.500 BIOT9T-0616.1 BIOT9T-0616.5 BIOT9T-0616.10
4-Nitrostyrol
4-Nitrostyrol
Formel: C₈H₇NO₂
MDL: MFCD00041254
CAS-Nummer: 100-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR