trans-β-Nitrostyrol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (E)-(2-Nitrovinyl)benzene, trans-1-Nitro-2-phenylethylene

L14367.06 L14367.14
L14367.06EA 29.1 EUR
L14367.06 L14367.14
trans-β-Nitrostyrol ≥98%
trans-β-Nitrostyrol

Purity: 98% CAS No.: 5153-67-3 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Storage Temperature: 2°C to 8°C Melting Point: 55-58° Boiling Point: 250-260° Beilstein Reference: 1210066 MDL No.: MFCD00007402

Formel: C₈H₇NO₂
Molecular Weight: 149.15 g/mol
Siedepunkt: 250…260 °C
Schmelzpunkt: 55…58 °C
MDL: MFCD00007402
CAS-Nummer: 5153-67-3
EINECS: 203-066-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR