trans-β-Nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-(2-Nitrovinyl)benzene, trans-1-Nitro-2-phenylethylene

M-Clarity™ Qualitätsstufe = MQ100

N26806-25G N26806-5G
SIALN26806-25GEA 110 EUR
SIALN26806-25G SIALN26806-5G
trans-β-Nitrostyrol, Sigma-Aldrich®
trans-β-Nitrostyrol
Formel: C₈H₇NO₂
Molecular Weight: 149,15 g/mol
MDL: MFCD00007402
CAS-Nummer: 5153-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR