β,4-Dinitrostyrol

Lieferant: Thermo Scientific

Synonyme: 1-nitro-4-(2-nitrovinyl)benzene

S11897.1 S11897.10 S11897.50
MAYBS11897.1EA 0 EUR
MAYBS11897.1 MAYBS11897.10 MAYBS11897.50
β,4-Dinitrostyrol
β,4-Dinitrostyrol
Formel: C₈H₆N₂O₄
MDL: MFCD00052058
CAS-Nummer: 3156-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR