3-Brom-5-chlor-4-iodbenzotrifluorid

Lieferant: Thermo Scientific

Synonyme: 1-Bromo-3-chloro-2-iodo-5-(trifluoromethyl)benzene

SEW01634.1 SEW01634.10
MAYBSEW01634.1EA 0 EUR
MAYBSEW01634.1 MAYBSEW01634.10
3-Brom-5-chlor-4-iodbenzotrifluorid
3-Brom-5-chlor-4-iodbenzotrifluorid
Formel: C₇H₂BrClF₃I
MDL: MFCD00042178
CAS-Nummer: 175205-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR